Please login to your online account to display your discounted pricing

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips


For water quality testing

Manufacturer: lamotte™  2979RSD

 View more versions of this product

Catalog No. 03-920-423

  • / Pack
 

Description & Specifications

Specifications

For Use With (Application) Water quality testing
Quantity 50 Strips per Vial
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Parameter Specifications Chlorine - Total
Range 0-0.5-1-3-5ppm
Product Type Single Factor Test Strips