Please login to your online account to display your discounted pricing

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips


For water quality testing

Manufacturer: lamotte™  3023GRSD

 View more versions of this product

Catalog No. 03-920-420

  • / Pack
 

Description & Specifications

Specifications

Range 0 to 6ppm
For Use With (Application) Water quality testing
Product Type Single Factor Test Strips
Parameter Specifications Ammonia
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Quantity 25 Strips per Vial