Please login to your online account to display your discounted pricing

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips


For water quality testing

Manufacturer: lamotte™  2996*

 View more versions of this product

Catalog No. 03-920-398

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Water quality testing
Parameter Specifications Nitrate and Nitrite
Quantity 50 Strips per Vial
Range 0-5-10-25-50 (nitrate), 0-0.5-1-5-10 (nitrite)
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Product Type Multi-Factor Test Strips