LaMotte™ DPD Chlorine Secondary Standards Kit for Series 1200 Colorimeters
Used for periodically rechecking calibration of Series 1200 Colorimeters
Supplier:  LaMotte™ LAM414003
Catalog No. 22-307-837
Includes: Blank and three standards, each with Certificate of Analysis, for low-, mid-, and high-range chlorine calibrations, packaged in a plastic case
Description
Description
  • Use once meter has been calibrated with a primary standard
  • Can be inserted periodically for a quick check of instrument calibration
Specifications
Specifications
Use once meter has been calibrated with a primary standard
Blank and three standards, each with certificate of analysis, for low, mid, and high-range chlorine calibrations, packaged in a plastic case
DPD Chlorine Secondary Standards Kit
Series 1200 colorimeter
Each