Please login to your online account to display your discounted pricing

LaMotte™ BioPaddles™ Dual-Agar Paddles for Food and Beverage


Contain microbe-specific media enclosed in a sterile vial

Manufacturer: lamotte™  5553/RMD

 View more versions of this product

Catalog No. 03-420-900

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 18-25°C
Sterile Yes
Quantity 40mL
Packaging Paddle
Media Type NUT-TTC/MAC dual Agar
Shelf Life 1 Year
Manufacturing Origin United States
Color Yellow
For Use With (Application) Food and beverage: isolation and differentiation of gram (-) enteric bacilli, Coliform testing/recovery of stressed Coliforms
Includes Free basic microbe identification application and access to technical information sheet
Accessories

1 Year