Please login to your online account to display your discounted pricing

Lamotrigine, Tocris

Inhibits glutamate release. Anticonvulsant

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Lamotrigine
Assay Percent Range >99%
Molecular Formula C9H7Cl2N5
Formula Weight 256.09
Synonym 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

161110

 
r & d systems
RDS161110
10mg Each for $177.37

161150

 
r & d systems
RDS161150
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 256.09
Quantity 10mg
Assay Percent Range >99%
Molecular Formula C9H7Cl2N5
CAS 84057-84-1
Synonym 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine
Chemical Name or Material Lamotrigine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature