missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Laminin Receptor Ab-1, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Immunofluorescence and Flow Cytometry, Thermo Scientific™ Laminin Receptor Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$234.82 - $1,360.92

Specifications

Antigen Laminin Receptor Ab-1
Clone MLuC5
Applications Immunofluorescence,Immunohistochemistry (Paraffin),Immunofluorescence,Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS259P Thermo Scientific™ Lab Vision™
MS259P
Purified Ab with BSA and Azide 1mL Each for $1,262.24
MS259P0 Thermo Scientific™ Lab Vision™
MS259P0
Purified Ab with BSA and Azide 100μL Each for $234.82
MS259P1 Thermo Scientific™ Lab Vision™
MS259P1
Purified Ab with BSA and Azide 500μL Each for $758.45
MS259P1ABX Thermo Scientific™ Lab Vision™
MS259P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $825.27
MS259PABX Thermo Scientific™ Lab Vision™
MS259PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,360.92
MS259R7 Thermo Scientific™ Lab Vision™
MS259R7
Ready-to-use for IHC (UVLP) 7mL Each for $652.86
Specifications

Description

Recommended for Immunohistochemistry (Formalin/paraffin), Immunofluorescence and Flow Cytometry, Thermo Scientific™ Laminin Receptor Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Laminin, one of the major glycoproteins of basement membrane, displays multiple biological activities which are mediated through its interaction with specific cell membrane receptors. Overexpression of laminin receptor is reportedly an independent prognostic factor in breast cancer and correlates with the dissemination of the tumor cells in bone marrow. Laminin receptor appears to play an important role in tumor invasion and metastasis.

Host Species: Mouse

Clone: MLuC5

Isotype: IgM

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Small cell lung carcinoma N592 live cells

Molecular Weight: 67kDa

Positive Control: Breast CA

Cellular Localization: Membrane with some cytoplasmic Ab-1 partially inhibits the binding of soluble laminin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry
  • Inhibits (partially) the binding of soluble laminin

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Laminin Receptor Ab-1
Immunofluorescence,Immunohistochemistry (Paraffin),Immunofluorescence,Immunohistochemistry (Paraffin)
Unlabeled
RUO
Primary
IgM
MLuC5
Monoclonal
Murine
Extracellular Matrix
Small cell lung carcinoma N592 live cells