missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Laminin B2/γ 1 Ab-2, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting (Not verified) and Immunoprecipitation (Not verified), Thermo Scientific™ Laminin B2/γ1 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

$265.94 - $1,184.41

Specifications

Antigen Laminin B2/gamma1 Ab-2
Clone D18
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1356P Thermo Scientific™ Lab Vision™
MS1356P
Purified Ab with BSA and Azide 1mL Each for $1,088.57
MS1356P0 Thermo Scientific™ Lab Vision™
MS1356P0
Purified Ab with BSA and Azide 100μL Each for $265.94
MS1356P1 Thermo Scientific
MS1356P1
Purified Ab with BSA and Azide 500μL Each for $694.16
MS1356P1ABX Thermo Scientific™ Lab Vision™
MS1356P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $761.33
MS1356PABX Thermo Scientific™ Lab Vision™
MS1356PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,184.41
MS1356R7 Thermo Scientific™ Lab Vision™
MS1356R7
Ready-to-use for IHC (UVLP) 7mL Each for $652.24
Specifications

Description

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting (Not verified) and Immunoprecipitation (Not verified), Thermo Scientific™ Laminin B2/γ1 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of α, β, and γ chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.

Host Species: Mouse

Clone: D18

Isotype: IgG2a/κ

Species Reactivity: Human, Rat and Guinea Pig. Others not known.

Epitope: Not determined

Immunogen: Rat glomerular basement membrane

Molecular Weight: ∽210kDa

Positive Control: LS174T cells, Skin

Cellular Localization: Basement membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunoprecipitation (Not verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Laminin B2/gamma1 Ab-2
Immunohistochemistry (Paraffin)
Unlabeled
RUO
Primary
IgG2a kappa
D18
Monoclonal
Murine
Cancer and Tumor Biology
Rat glomerular basement membrane