Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Laminin B2/γ 1 Ab-2, Mouse Monoclonal Antibody


Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting (Not verified) and Immunoprecipitation (Not verified), Thermo Scientific™ Laminin B2/γ1 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS1356PABX

 View more versions of this product

Catalog No. MS1356PABX

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2a kappa
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Rat
Quantity 200µL
Antigen Laminin B2/gamma1 Ab-2
Immunogen Rat glomerular basement membrane
Clone D18
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Format Concentrated
Host Species Mouse
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Classification Monoclonal
Formulation Ab without BSA/Azide (Frozen)
Purification Method LS174T cells, Skin
Research Discipline Cancer and Tumor Biology

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting (Not verified) and Immunoprecipitation (Not verified), Thermo Scientific™ Laminin B2/γ1 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of α, β, and γ chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.

Host Species: Mouse

Clone: D18

Isotype: IgG2a/κ

Species Reactivity: Human, Rat and Guinea Pig. Others not known.

Epitope: Not determined

Immunogen: Rat glomerular basement membrane

Molecular Weight: ∽210kDa

Positive Control: LS174T cells, Skin

Cellular Localization: Basement membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunoprecipitation (Not verified)

Note:

USA: RUO; Int'l: RUO