missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Laminin B1/β 1 Ab-1, Rat Monoclonal Antibody

Recommended for a variety of molecular biology techniques, Thermo Scientific™ Laminin B1/β1 Ab-1, Rat Monoclonal Antibody provides accurate, reproducible results.

$277.78 - $1,463.41

Specifications

Antigen Laminin B1/beta1 Ab-1
Clone LT3
Applications Immunohistochemistry (Frozen),Immunoprecipitation,Western Blotting,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blotting,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blotting
Classification Monoclonal
Format Concentrated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
RT796B Thermo Scientific™ Lab Vision™
RT796B
200µg/mL Biotin 1mL Each for $1,463.41
RT796B0 Thermo Scientific™ Lab Vision™
RT796B0
200µg/mL Biotin 100μL Each for $372.16
RT796B1 Thermo Scientific™ Lab Vision™
RT796B1
200µg/mL Biotin 500μL Each for $925.10
RT796P Thermo Scientific™ Lab Vision™
RT796P
200µg/mL Unlabeled 1mL Each for $1,123.18
RT796P0 Thermo Scientific™ Lab Vision™
RT796P0
200µg/mL Unlabeled 100μL Each for $277.78
RT796P1 Thermo Scientific™ Lab Vision™
RT796P1
200µg/mL Unlabeled 500μL Each for $713.39
RT796P1ABX Thermo Scientific™ Lab Vision™
RT796P1ABX
1mg/mL Unlabeled 100μL Each for $808.53
RT796PABX Thermo Scientific™ Lab Vision™
RT796PABX
1mg/mL Unlabeled 200μL Each for $1,221.80
Specifications

Description

Recommended for a variety of molecular biology techniques, Thermo Scientific™ Laminin B1/β1 Ab-1, Rat Monoclonal Antibody provides accurate, reproducible results.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of α, β, and γ chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.

Host Species: Rat

Clone: LT3

Isotype: IgG1/κ

Species Reactivity: Human, Pig, Hamster, and Mouse. Others not known.

Epitope: Not determined

Immunogen: Murine EHS laminin preparation

Molecular Weight: ∽210kDa

Positive Control: LS174T cells, Normal colon or colon CA

Cellular Localization: Basement membrane Ab-1 reacts with laminin B chain. It stains most human and murine basement membranes, except for glomerular and arterial ones.

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen and alcohol-fixed, paraffin-embedded)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Laminin B1/beta1 Ab-1
Immunohistochemistry (Frozen),Immunoprecipitation,Western Blotting,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blotting,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blotting
Concentrated
RUO
Primary
IgG2a kappa
LT3
Monoclonal
Rat
Extracellular Matrix
Murine EHS laminin preparation