Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Laminin B1/β 1 Ab-1, Rat Monoclonal Antibody


Recommended for a variety of molecular biology techniques, Thermo Scientific™ Laminin B1/β1 Ab-1, Rat Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  RT796P1ABX

 View more versions of this product

Catalog No. RT796P1ABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Clone LT3
Isotype IgG2a kappa
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 100µL
Immunogen Murine EHS laminin preparation
Antigen Laminin B1/beta1 Ab-1
Host Species Rat
Regulatory Status RUO
Conjugate Unlabeled
Research Discipline Extracellular Matrix
Type Primary
Classification Monoclonal
Concentration 1mg/mL

Recommended for a variety of molecular biology techniques, Thermo Scientific™ Laminin B1/β1 Ab-1, Rat Monoclonal Antibody provides accurate, reproducible results.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of α, β, and γ chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.

Host Species: Rat

Clone: LT3

Isotype: IgG1/κ

Species Reactivity: Human, Pig, Hamster, and Mouse. Others not known.

Epitope: Not determined

Immunogen: Murine EHS laminin preparation

Molecular Weight: ∽210kDa

Positive Control: LS174T cells, Normal colon or colon CA

Cellular Localization: Basement membrane Ab-1 reacts with laminin B chain. It stains most human and murine basement membranes, except for glomerular and arterial ones.

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen and alcohol-fixed, paraffin-embedded)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO