Please login to your online account to display your discounted pricing

Alfa Aesar™ Laminin (925-933)

For cellular and molecular biology applications

Specifications

Product Type Laminin (925-933)
Molecular Weight 967.05
Storage Requirements -30° to -10°C
Sequence Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg
For Use With (Application) Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64340EXD

 
alfa aesar™
J64340EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ64340MCR

 
alfa aesar™
J64340MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Laminin (925-933)
Molecular Weight 967.05
Storage Requirements -30° to -10°C
Sequence Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg
For Use With (Application) Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor
Format Solid
Regulatory Status RUO
Purification Method HPLC