missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Laminarin, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6619303

 View more versions of this product

Catalog No. AAJ6619303



Description

Description

Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Laminarin is a storage glucan and is now used to study it’s interaction with laminarinase as its substrate.

Solubility
Soluble in dimethyl sulfoxide (slightly, heated). Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Laminarin
White
MFCD00046691
14,5350
Soluble in dimethyl sulfoxide (slightly,heated). Slightly soluble in water.
−45° (c=2 in Water)
(2S,3R,4S,5R,6R)-4-[(2S,3R,4S,5R,6R)-3,5-dihydroxy-6-(hydroxymethyl)-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,5-triol
46173707
9008-22-4
C18H32O16
1 g
laminarin, laminaran
DBTMGCOVALSLOR-DEVYUCJPSA-N
C(C1C(C(C(C(O1)O)O)OC2C(C(C(C(O2)CO)O)OC3C(C(C(C(O3)CO)O)O)O)O)O)O
504.438
Powder

Chemical Identifiers

9008-22-4
504.438
DBTMGCOVALSLOR-DEVYUCJPSA-N
46173707
C(C1C(C(C(C(O1)O)O)OC2C(C(C(C(O2)CO)O)OC3C(C(C(C(O3)CO)O)O)O)O)O)O
C18H32O16
MFCD00046691
laminarin, laminaran
(2S,3R,4S,5R,6R)-4-[(2S,3R,4S,5R,6R)-3,5-dihydroxy-6-(hydroxymethyl)-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,5-triol
Safety and Handling

Safety and Handling

EINECSNumber : 232-712-4

RTECSNumber : OE1523500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only