Please login to your online account to display your discounted pricing

Lamin A/C Mouse, Clone: 4A7, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $146.45

Specifications

Antigen Lamin A/C
Clone 4A7
Host Species Murine
Gene Alias LMNA, LMNC
Species Reactivity Hamster, Human, Mouse, Rabbit
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122797

 
life technologies
LS14968880
25μg Each for $146.45
Description & Specifications

Specifications

Antigen Lamin A/C
Clone 4A7
Host Species Murine
Gene Alias LMNA, LMNC
Species Reactivity Hamster, Human, Mouse, Rabbit
Applications ELISA
Applications Immunocytochemistry
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 4A7 recognizes human, mouse, rabbit and hamster lamin A/C. Lamins are nuclear intermediate filament proteins that provide framework for the nuclear envelope, maintain cell morphology, and protect the nucleus from mechanical, thermal, and oxidative stresses. Lamins also play a role in nuclear assembly, chromatin organization, DNA replication, RNA transcription, cell signaling, and apoptosis. Lamin C is a splice variant of Lamin A and both are A-type lamins. Lamin A and C are crucial for skeletal and cardiac development and function. Defects in A-type lamins result in cardiomyopathy, muscular dystrophy, peripheral neuropathy, lipodystrophy, restrictive dermopathy, and progeroid disorders.