Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody provides accurate, reproducible results.

$242.45 - $770.57

Specifications

Antigen Lambda Light Chain Ab-4 (Cocktail)
Clone LAM03 + HP6054
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1158P

 
thermo scientific™ lab vision™
MS1158P
Purified Ab with BSA and Azide 1mL Each for $675.88

MS1158P0

 
thermo scientific™ lab vision™
MS1158P0
Purified Ab with BSA and Azide 100μL Each for $242.45

MS1158P1

 
thermo scientific™ lab vision™
MS1158P1
Purified Ab with BSA and Azide 500μL Each for $478.37

MS1158P1ABX

 
thermo scientific™ lab vision™
MS1158P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $607.73

MS1158PABX

 
thermo scientific™ lab vision™
MS1158PABX
Ab without BSA/Azide (Frozen) 200μL Each for $770.57

MS1158R7

 
thermo scientific™ lab vision™
MS1158R7
Ready-to-use for IHC (UVLP) 7mL Each for $351.61
Description & Specifications

Specifications

Antigen Lambda Light Chain Ab-4 (Cocktail)
Clone LAM03 + HP6054
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Immunogen Purified human lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads
Isotype IgG2a kappa

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody provides accurate, reproducible results.

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: LAM03 + HP6054

Isotype: IgG1/κ + IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human Lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic Ab-4 cocktail is especially designed for sensitive detection of lambda light chain of immunoglobulin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO