Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody


Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS1158P1ABX

 View more versions of this product

Catalog No. MS1158P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Quantity 100µL
Antigen Lambda Light Chain Ab-4 (Cocktail)
Isotype IgG2a kappa
Cross Reactivity Human
Immunogen Purified human lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads
Format Concentrated
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Clone LAM03 + HP6054
Classification Monoclonal
Type Primary
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Concentration 1mg/mL
Research Discipline Hematopoietic
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ Lambda Light Chain Ab-4 (Cocktail), Mouse Monoclonal Antibody provides accurate, reproducible results.

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: LAM03 + HP6054

Isotype: IgG1/κ + IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human Lambda light chain, and purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic Ab-4 cocktail is especially designed for sensitive detection of lambda light chain of immunoglobulin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO