missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lakeside™ Standard-Duty Utility Cart

A Lakeside Standard-Duty Utility Cart is ideal for light-duty transport and storage in the laboratory.

Manufacturer:  Lakeside™ LKM311

 View more versions of this product

Catalog No. 11-954-750



Description

Description

A Lakeside Standard-Duty Utility Cart is ideal for light-duty transport and storage in the laboratory.

  • Load capacity: 300 lb. (136kg)
  • Durable welded stainless-steel construction cleans and sanitizes easily
  • Designed for continuous transport even over uneven floor surfaces
  • Integrated push handle for easy handling
  • Three fixed, open shelves with reinforced, hemmed front for added strength
  • Sound-deadening panels under shelves restrict vibrations and noise
  • Bumpers on legs and handles protect walls and furniture
  • Four 3 1/2 in. dia. (8.9cm) rubber-cushioned, swivel casters for maneuverability

Order Info

Shipped assembled. Weight: 29 lb. (13.2kg)

Specifications

Specifications

32.13 in.
300 lb.
Stainless Steel
27.5 in.
41.2cm
3
Integrated push handle, 24 L x 15.5 in. W three fixed shelves with reinforced hemmed front, 3.5 in. dia. four Rubber cushioned, swivel casters, leg and handle bumpers, sound deadening panels
16.25 in.
136 kg.
81.61cm
69.85cm
27.5 x 16.25 x 32.13 in. (69.85 x 41.2 x 81.61cm)
Ideal for dining area bussing, set up, tray or soiled dish station, light duty transport and storage and continuous transporting over even floor surfaces
SDS
Documents