missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lakeside™ Medium-Duty Utility Cart DFS Item

Lakeside Medium-Duty Utility Cart ideal for moderate weight loads and all-purpose back-of-the-house duties.

Manufacturer:  Lakeside™ LKM444

 View more versions of this product

Catalog No. 11-954-758



Description

Description

  • Load capacity: 500 lb. (230kg)
  • 20 gauge shelves; reinforced top-shelf
  • Unitized, electronically-welded, 16-gage angle leg frame
  • Protective bumpers on legs and handle
  • Sound deadening panels under shelves restrict vibrations and noise

Specifications

Specifications

90 cm
Medium-duty Utility Cart
Ideal for moderate weight loads and all purpose back of the house duties as well as constant use over various standard floor surfaces
94.6 cm
39.25 in.
500 lb.
Stainless Steel
22.36 in.; Shelf: 21 in.
53.34 cm
39.25 x 22.36 x 37.25 in. (99.7 x 56.8 x 94.6 cm)
37.25 in.
16G Angle leg frame, 21 x 35 in. W three shelves, legs and handle bumpers, sound deadening panels, 5 in. dia. extra load cushion casters
100 cm
230 kg
3
56.8 cm
SDS
Documents
Product Certifications
Promotions

Promotions