Please login to your online account to display your discounted pricing

Lactic acid, lithium salt, 99%, ACROS Organics™

$35.36 - $100.98

Chemical Identifiers

Chemical Name or Material Lactic acid, lithium salt
CAS 867-55-0
Purity Grade Extra Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3H5LiO3
Linear Formula CH3CH(OH)CO2Li
Formula Weight 96.01
MDL Number MFCD00065399
Synonym Lactic acid lithium salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC413335000

  SDS
thermo fisher scientific
AC413335000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC413331000

  SDS
thermo fisher scientific
AC413331000
Glass bottle 100g Each for $100.98

AC413330250

  SDS
fisher scientific acros
413330250
Glass bottle 25g Each for $35.36

AC413330050

  SDS
fisher scientific acros
413330050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Purity Grade Extra Pure
Physical Form Crystalline Powder
Formula Weight 96.01
Quantity 500g
Color White
pH 6 to 9 (1% soln.)
Beilstein 03, IV, 633
Absorbance (10% aq. soln.)
Synonym Lactic acid lithium salt
Chemical Name or Material Lactic acid, lithium salt
Packaging Glass bottle
Molecular Formula C3H5LiO3
CAS 867-55-0
Assay Percent Range 99%
MDL Number MFCD00065399
Appearance of Solution (10% aq. soln.) clear, colorless
Absorbance Conditions (10% aq. soln.)
Linear Formula CH3CH(OH)CO2Li
Infrared Spectrum Authentic