Please login to your online account to display your discounted pricing

Lactic acid, lithium salt, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  413330250

 View more versions of this product

Catalog No. AC413330250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Purity Grade Extra Pure
Physical Form Crystalline Powder
Formula Weight 96.01
Quantity 25g
Color White
Beilstein 03, IV, 633
pH 6 to 9 (1% soln.)
Absorbance (10% aq. soln.)
Absorbance Conditions (10% aq. soln.)
Assay Percent Range 99%
Molecular Formula C3H5LiO3
Linear Formula CH3CH(OH)CO2Li
Chemical Name or Material Lactic acid, lithium salt
MDL Number MFCD00065399
CAS 867-55-0
Appearance of Solution (10% aq. soln.) clear, colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Lactic acid lithium salt

Specifications

Chemical Name or Material Lactic acid, lithium salt
CAS 867-55-0
Purity Grade Extra Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3H5LiO3
Linear Formula CH3CH(OH)CO2Li
Formula Weight 96.01
MDL Number MFCD00065399
Synonym Lactic acid lithium salt