Please login to your online account to display your discounted pricing

Lactic Acid, Certified, 10.0% (v/v) ±0.5%, LabChem™

$69.77 - $69.77

Chemical Identifiers

Chemical Name or Material Lactic Acid
CAS 7732-18-5
CAS Max % 90.19
CAS 50-21-5
CAS Max % 9.81
Purity Grade Certified Grade
Linear Formula C3H6O3
Formula Weight 90.08
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC160001

 
lab chem
LC160001
500mL Each for $69.77
Description & Specifications

Specifications

Formula Weight 90.08
Assay 10.0% ± 0.5% (v/v)
Purity Grade Certified Grade
CAS Max % 9.81
Decomposition Information Carbon monoxide; Carbon dioxide
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Lactic Acid
Identification Passes Test
Traceability to NIST Traceable to NIST
CAS 7732-18-5
Color Clear, colorless
Packaging Poly Bottle
Recommended Storage Room Temperature
CAS Max % 90.19
Linear Formula C3H6O3
Density 1.02g/mL
Specific Gravity 1.02g/mL
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Solubility Soluble in water
CAS 50-21-5
Physical Form Liquid
Quantity 500mL
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

Room Temperature