missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lactic Acid, ACS Grade, 85.0 to 90.0%, LabChem™

$124.03 - $124.03

Chemical Identifiers

CAS 50-21-5,7732-18-5
CAS Max % 85,15
Linear Formula C3H6O3
Formula Weight 90.08
UN Number UN3265
Chemical Name or Material Lactic Acid
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC159501 Lab Chem
LC159501
500mL Each for $124.03
Specifications Description & Specifications

Chemical Identifiers

50-21-5,7732-18-5
C3H6O3
UN3265
ACS Grade
85,15
90.08
Lactic Acid

Specifications

50-21-5,7732-18-5
Liquid
90.08
Passes Test
Soluble in water
16.8°C
122°C
113°C
85.0 to 90.0%
85,15
C3H6O3
UN3265
Amber Glass
Carbon monoxide; Carbon dioxide
Clear, colorless
500mL
Lactic Acid
ACS Grade

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.

GHS P Statement
Keep only in original container.
Do not breathe mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 200-018-0

Recommended Storage : Room Temperature