missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lacidipine, TCI America™

Manufacturer:  TCI America L0276200MG

 View more versions of this product

Catalog No. L0276200MG



Specifications

Specifications

Lacidipine
179°C
103890-78-4
MFCD00865936
CCOC(=O)C1=C(NC(=C(C1C2=CC=CC=C2C=CC(=O)OC(C)(C)C)C(=O)OCC)C)C
455.551
455.55
≥98.0% (HPLC,N)
White-Yellow
200mg
C26H33NO6
GKQPCPXONLDCMU-CCEZHUSRSA-N
diethyl 2,6-dimethyl-4-[2-[(E)-3-[(2-methylpropan-2-yl)oxy]-3-oxoprop-1-enyl]phenyl]-1,4-dihydropyridine-3,5-dicarboxylate
5311217
Crystal-Powder at 20°C

Chemical Identifiers

103890-78-4
455.551
GKQPCPXONLDCMU-CCEZHUSRSA-N
diethyl 2,6-dimethyl-4-[2-[(E)-3-[(2-methylpropan-2-yl)oxy]-3-oxoprop-1-enyl]phenyl]-1,4-dihydropyridine-3,5-dicarboxylate
C26H33NO6
MFCD00865936
5311217
CCOC(=O)C1=C(NC(=C(C1C2=CC=CC=C2C=CC(=O)OC(C)(C)C)C(=O)OCC)C)C
Safety and Handling

Safety and Handling

RTECSNumber : US7970200

TSCA : No

Recommended Storage : Refrigerator

Documents