missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lacidipine, TCI America™

$238.34 - $832.66

Chemical Identifiers

CAS 103890-78-4
Molecular Formula C26H33NO6
Molecular Weight (g/mol) 455.551
MDL Number MFCD00865936
InChI Key GKQPCPXONLDCMU-CCEZHUSRSA-N
PubChem CID 5311217
IUPAC Name diethyl 2,6-dimethyl-4-[2-[(E)-3-[(2-methylpropan-2-yl)oxy]-3-oxoprop-1-enyl]phenyl]-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C2=CC=CC=C2C=CC(=O)OC(C)(C)C)C(=O)OCC)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L0276200MG View Documents TCI America
L0276200MG
200mg Each for $238.34
 
L02761G View Documents TCI America
L02761G
1g Each for $832.66
 
Specifications

Chemical Identifiers

103890-78-4
455.551
GKQPCPXONLDCMU-CCEZHUSRSA-N
diethyl 2,6-dimethyl-4-[2-[(E)-3-[(2-methylpropan-2-yl)oxy]-3-oxoprop-1-enyl]phenyl]-1,4-dihydropyridine-3,5-dicarboxylate
C26H33NO6
MFCD00865936
5311217
CCOC(=O)C1=C(NC(=C(C1C2=CC=CC=C2C=CC(=O)OC(C)(C)C)C(=O)OCC)C)C

Specifications

White-Yellow
200mg
C26H33NO6
GKQPCPXONLDCMU-CCEZHUSRSA-N
diethyl 2,6-dimethyl-4-[2-[(E)-3-[(2-methylpropan-2-yl)oxy]-3-oxoprop-1-enyl]phenyl]-1,4-dihydropyridine-3,5-dicarboxylate
5311217
Crystal-Powder at 20°C
Lacidipine
179°C
103890-78-4
MFCD00865936
CCOC(=O)C1=C(NC(=C(C1C2=CC=CC=C2C=CC(=O)OC(C)(C)C)C(=O)OCC)C)C
455.551
455.55
≥98.0% (HPLC,N)
Safety and Handling

Safety and Handling

RTECSNumber : US7970200

TSCA : No

Recommended Storage : Refrigerator