missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Labtainer™ BPC Bag with 3 Ports, MPC Insert and Injection Port

Simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

$74.88 - $152.11

Specifications

Certifications/Compliance cGMP
Description Two-dimensional disposable cell culture/bioprocess bag
Film Description CX5-14 Single Web Film
Port Detail 3/8in. Quick Disconnect Insert; 3/8in. Quick Disconnect Insert; 1/4in. Injection Port
Sterilization Method Gamma Irradiated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
SH3070901 Thermo Scientific™
SH3070901
5L Pack for $77.66
SH3070905 Thermo Scientific™
SH30709.05
2L Each for $74.88
SH3070903 Thermo Scientific™
SH3070903
20L Pack for $93.32
SH3070902 Thermo Scientific™
SH3070902
10L Pack for $87.09
SH3070904 Thermo Scientific™
SH3070904
50L Pack for $152.11
Specifications

Description

Using Thermo Scientific™ Labtainer™ BPC bag with 3 ports (MPC insert and injection), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioproces liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation system

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Description & Specifications

Specifications

cGMP
CX5-14 Single Web Film
Gamma Irradiated
No
Two-dimensional disposable cell culture/bioprocess bag
3/8in. Quick Disconnect Insert; 3/8in. Quick Disconnect Insert; 1/4in. Injection Port
Ambient