missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Labtainer™ BPC Bag with 3 Ports, Luer Lock

Simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

$80.97 - $158.35

Specifications

Certifications/Compliance cGMP
Description Two-dimensional disposable cell culture/bioprocess bag
Film Description CX5-14 Single Web Film
Port Detail Female 1/8” Luer Connector, Male 3/8” MPC Connector, Female 3/8” MPC Connector
Sterilization Method Gamma Irradiated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
SH3071301 Thermo Scientific™
SH3071301
5L Pack for $90.05
SH3071302 Thermo Scientific™
SH3071302
10L Pack for $100.54
SH3071303 Thermo Scientific™
SH3071303
20L Pack for $100.54
SH3071304 Thermo Scientific™
SH3071304
50L Pack for $158.35
SH3071305 Thermo Scientific™
SH3071305
2L Each for $80.97
Specifications

Description

Using Thermo Scientific™ Labtainer™ BPC bag with 3 ports (luer lock), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.

Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.

Highlights:

  • 11 sizes from 50mL to 50L
  • Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Convenient end-port design with a handle at the opposing end
  • Space-efficient, ergonomic alternative to rigid walled carboys
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Sampling – collection of in-process samples at a variety of bioprocessing stages

  • Harvest – collection and storage of bulk product from bioreactors

  • Storage – sterile storage of media, buffers and process intermediates

  • Separation – feeding and receiving of sterile liquids from separation systems

  • Purification – buffer feeding to purification system, fraction collection and storage

  • Filling – sterile reservoir for filling systems

  • Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Description & Specifications

Specifications

cGMP
CX5-14 Single Web Film
Gamma Irradiated
No
Two-dimensional disposable cell culture/bioprocess bag
Female 1/8” Luer Connector, Male 3/8” MPC Connector, Female 3/8” MPC Connector
Ambient