Labplas Twirl'EM™ Sterile Sampling Bags, SafetyTabs
Provide a sterile, secure, contaminant-free pliant container that ensures dependable analysis results. Labplas Twirl'EM™ Sterile Sampling Bags with safety tabs incorporate easy-to-pull tabs that open the bag without risk of contaminating the bag interior. They are useful in a variety of sampling applications, including environmental and surface sampling.
$329.69 - $719.31
Specifications
Material PE (Polyethylene)
View More Specs
Products 10
Catalog Number Mfr. No. Capacity (Metric) Closure Type Description Thickness (English) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Type Description Thickness (English) Price Quantity  
01-815-304 View Documents Labplas
EPL4590
450mL Closure with two round wires Clear body 2.5 mil Case of 1000 Each for $329.69
01-815-308 View Documents Labplas
EPL7012
1650mL Closure with two round wires Clear body 3 mil Case of 1000 Each for $653.64
01815309 View Documents Labplas
EPL47012
1650mL Closure with two round wires Clear body 4 mil Case of 1000 Each for $699.14
01-815-315 View Documents Labplas
EPR5590
650mL Closure with two round wires Printed body 3 mil Case of 1000 Each for $455.00
01-815-314 View Documents Labplas
EPR4590
450mL Closure with two round wires Printed body 2.5 mil Case of 1000 Each for $416.38
01-815-317 View Documents Labplas
EPR7012
1650mL Closure with two round wires Printed body 3 mil Case of 1000 Each for $602.53
31-551-06 View Documents Labplas
EFR3070
150mL Closure with one round and one flat wire Printed body 2.5 mil Case of 1000 Each for $365.83
31-551-05 View Documents Labplas
EFL5512
1080mL Closure with one round and one flat wire Clear body 3 mil Case of 1000 Each for $719.31
31-551-04 View Documents Labplas
EFR4590
450mL Closure with one round and one flat wire Printed body 2.5 mil Case of 1000 Each for $457.53
31-551-08 View Documents Labplas
EFR5590
650mL Closure with one round and one flat wire Printed body 3 mil Case of 1000 Each for $595.97
Description
Description
  • Virgin polyethylene tubing
  • Can be heated to 240°C for sterility
  • Large bag mouth opening to facilite insertion of samples
  • Easy pull tabs to open and close the bag with decreased risk of contamination
  • R-Nase, D-Nase, Pyrogen, and BPA free
Applications include:
  • Enviromental sampling
  • Surface sampling
  • Carcass sampling
  • Biomedical research
  • Pharmaceutical research
  • Food industry
Certifications
Sterility
Compliance
EPA, FDA, USDA, HACCP
Specifications
Specifications
PE (Polyethylene)