Please login to your online account to display your discounted pricing
Labplas Twirl'EM™ Large Format Sampling Bags with Tabs
Polyethylene sampling bags with large openings
Manufacturer: labplas inc  BDL41218
 View all options for this product
Catalog No. 01-815-366
Description & Specifications
Specifications
Zip Close Regular tabs
Material Polyethylene
Capacity (Metric) 6750mL
Thickness 4 mil
Sterile Yes
Product Style Regular Tabs
  • Large sized
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs