Labplas Filtra-Bag Blender Bags
Designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances. Labplas Filtra-Bag Blender Bags are constructed with a polyethylene outer shell that reduces the risk of piercing the bag wall during blending.
$155.72 - $1,437.86
Specifications
For Use With (Application) Aliquoting
Material PE (Polyethylene)
Sterility Sterile
Thickness (English) 3 mil
View More Specs
Products 8
Catalog Number Mfr. No. Capacity (Metric) Closure Type Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Type Price Quantity  
01-002-57
View Documents
Labplas
SCTO7012A
1650mL Open Top
Pack of 100 Each for $155.72
Case of 4 Pack for $622.88
01-815-386
View Documents
Labplas
SCT6090A
710mL Tear-off protection strip
Case of 400 Each for $607.67
01-815-387
View Documents
Labplas
SCT7012A
1650mL Tear-off protection strip
Case of 400 Each for $606.86
01-815-389
View Documents
Labplas
SCT1520A
12000mL Tear-off protection strip
Case of 200 Each for $776.92
01-815-390
View Documents
Labplas
EFT6090A
710mL Safety tabs
Case of 400 Each for $614.86
01-815-391
View Documents
Labplas
EFT7012A
1650 mL Safety tabs
Case of 400 Each for $850.56
01-815-392
View Documents
Labplas
EFT1015A
2720mL Safety tabs
Case of 400 Each for $1,437.86
01-815-376
View Documents
Labplas
EFT1515A
4000mL Safety tabs
Case of 200 Each for $810.39
Description
Description
Designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid/solid substances. Labplas Filtra-Bag Blender Bags are constructed with a polyethylene outer shell that reduces the risk of piercing the bag wall during blending.
  • Used for aliquoting
  • Blender accessible
  • Filter membrane for separating solids and liquids
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
Specifications
Specifications
Aliquoting
PE (Polyethylene)
Sterile
3 mil
SDS