missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fischer Technical Company™ Laboratory Silicone Safety Mat

Silicone Safety Mats provide protection for your lab station from spills and slippage.

Manufacturer:  Fischer Technical Company™ MATRED

 View more versions of this product

Catalog No. S64978



Description

Description

Laboratory Silicone Safety Mat is designed to protect your desktop or lab station and prevent glassware from sliding

  • Available in red or black
  • Large mat is 350 x 350 mm
  • Small mat is 250 x 250 mm
  • Silicone constructions provides protection against many chemicals
Specifications

Specifications

Lab Mat
9.8 in.
9.8 in.
250 x 250 mm (9.8 x 9.8 in.)
Silicone
250 mm
250 mm
Red
SDS
Documents
Product Certifications
Promotions

Promotions