missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Laboratory Performance Check Standard without Mercury For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC LPC1500N
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
K | 100μg/mL |
P | 100μg/mL |
SiO&subscript_2; | 100μg/mL |
Al | 20μg/mL |
As | 20μg/mL |
B | 20μg/mL |
Ba | 20μg/mL |
Be | 20μg/mL |
Ca | 20μg/mL |
Cd | 20μg/mL |
Co | 20μg/mL |
Cr | 20μg/mL |
Cu | 20μg/mL |
Fe | 20μg/mL |
Li | 20μg/mL |
Mg | 20μg/mL |
Mn | 20μg/mL |
Mo | 20μg/mL |
Na | 20μg/mL |
Ni | 20μg/mL |
Pb | 20μg/mL |
Sb | 20μg/mL |
Se | 20μg/mL |
Sn | 20μg/mL |
Sr | 20μg/mL |
Tl | 20μg/mL |
V | 20μg/mL |
Zn | 20μg/mL |
Ag | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
LPC Standard 1 without Mercury | |
500 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Various |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Environmental | |
5% HNO3/tr Tart. Acid/tr HF |
Product intended for laboratory use only