missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Laboratory Fortifying Stock Solution For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC LFS1100
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
P | 50μg/mL |
Al | 25μg/mL |
As | 25μg/mL |
B | 25μg/mL |
Ba | 25μg/mL |
Cr | 25μg/mL |
Cu | 25μg/mL |
Fe | 25μg/mL |
Li | 25μg/mL |
Mn | 25μg/mL |
Ni | 25μg/mL |
Pb | 25μg/mL |
Sb | 25μg/mL |
Se | 25μg/mL |
SiO&subscript_2; | 25μg/mL |
Sr | 25μg/mL |
Tl | 25μg/mL |
Zn | 25μg/mL |
Cd | 10μg/mL |
Co | 10μg/mL |
Mo | 10μg/mL |
Sn | 10μg/mL |
V | 10μg/mL |
Be | 5μg/mL |
Ag | 2.5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
LFS Solution 1 | |
125 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Environmental | |
5% HNO3/tr Tart. Acid/tr HF |
Triple-leached LDPE bottle | |
With Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Components in 5% HNO3/tr. Tartaric acid/tr. HF. Mercury is supplied separately. Al •25μg/mL Sb •25μg/mL As •25μg/mL Ba •25μg/mL Be •5μg/mL B •25μg/mL Cd •10μg/mL Cr •25μg/mL Co •10μg/mL Cu •25μg/mL Fe •25μg/mL Pb •25μg/mL Li •25μg/mL Mn •25μg/mL Hg •5μg/mL Mo •10μg/mL Ni •25μg/mL P •50μg/mL Se •25μg/mL SiO2 |
Product intended for laboratory use only