Please login to your online account to display your discounted pricing

Laboratory Charts

Instant access to common laboratory techniques and safety rules.

$140.40 - $359.10

Specifications

Classification Science
For Use With (Application) Lists 23 important safety rules or 23 common laboratory techniques
View More Specs
Products
Catalog Number Mfr. No. Product Type Quantity Price Quantity    

S45572

 
fisher science education
2028-10EMD
Laboratory Techniques Chart 1/Ea. Each for $140.40

S45571

 
fisher science education
2027-10EMD
Laboratory Safety Chart 1/Ea. Each for $140.40

S65021

 
fisher science education
202812
Laboratory Techniques Chart 3/Pk. Pack of 3 Each for $359.10

S65022

 
fisher science education
202712
Laboratory Safety Chart 3/Pk. Pack of 3 Each for $359.10
Description & Specifications

Specifications

Classification Science
For Use With (Application) Lists 23 important safety rules or 23 common laboratory techniques

  • List 23 important safety rules or 23 common laboratory techniques
  • Printed on tear-proof synthetic stock with metal strips for hanging
  • Dimensions: 36 × 44 in. (91.4 × 111.8cm)