Please login to your online account to display your discounted pricing

Laboratory Charts


Instant access to common laboratory techniques and safety rules.

Manufacturer: fisher science education  202812

 View more versions of this product

Catalog No. S65021

  • / Pack of 3 Each
 

Description & Specifications

Specifications

Product Type Laboratory Techniques Chart
Classification Science
For Use With (Application) Lists 23 important safety rules or 23 common laboratory techniques
Quantity 3/Pk.