LW Series 40X Objective
Replacement objective
Manufacturer:  Fisher Scientific 7123040
Catalog No. S96064
Description
Description
  • NA 0.6/WD 2.25mm with phase slider (LW series Phase Plan)²
Specifications
Specifications
Education
LW Series
40X
Documents