Learn More
Brady™ Labels Vinyl HIGH VOLTAGE
Catalog No. 17108802
Change view
Click to view available options
Dimensions (L x W):
3.5 x 5 in.
Catalog No. Dimensions (L x W)
17108802 3.5 x 5 in.
1 options
Catalog No. 17108802 Supplier Brady™ Supplier No. 20990LSA
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Clearly mark high-voltage hazards with durable labels designed for long-lasting visibility.
  • High-performance, pressure-sensitive adhesive labels withstand up to years of use
  • Good resistance to most common chemicals, oils and water
  • Service temperature of -40°F to +122°F (-40°C to +50°C)
Specifications
No. per Pack 20 Pk.
Dimensions (L x W) 3.5 x 5 in.