Labconco™ XPert™ Bulk Powder Filtered Systems
Protects from hazardous airborne particulates and fumes generated during powder transfer operations
Supplier: Labconco™ 3961601
Description
- Clean-Sweep™ openings and a zone-perforated rear baffle enhance horizontal laminar airflow
- With built-in Guardian™ 1000 digital airflow monitor
- Upper dilution air supply with by-pass air
- Upper containment sash foil and side-entry air foils
- Stainless steel work surface with front spill trough
- Transfer caddy with caddy latch and positioning plate
- Push-button operated electric hydraulic lift mechanism for raising the container to the optimal ergonomic height
- 30 in. interior depth and 32 in. interior height
- Tempered safety glass front sash and sides
- 12 in. high sash opening; ergonomic angled sash with sealed gas-assisted springs hold sash open (28 in. loading height)
- Built-in blower; outside ducting not required
- 99.99% efficient HEPA filter
- True bag-in/bag-out filter disposal system
- Pre-wired fluorescent light and switches for remote blower and light
- Epoxy-coated steel exterior
- 2 in. square-tube epoxy-coated steel base stand
- Left-side mounted waste chute
- Fourplex electrical receptacle and two utility ports
Compliance
SEFA 1-2002, Modified ASHRAE™ 110-1995, ANSI™ Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1
Specifications
Fume Hood | |
SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE | |
93.98 cm | |
12 in. | |
1105 lb. |
Fluorescent | |
37 in. | |
83 in. | |
30.5 cm | |
182.8 cm |