Please login to your online account to display your discounted pricing

Labconco™ XPert™ Bulk Powder Filtered Systems


Protects from hazardous airborne particulates and fumes generated during powder transfer operations

Manufacturer: labconco™  LCN3961500

 View more versions of this product

Catalog No. 10-265-140

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Sash Opening (Metric) 30.5cm
Width (English) 60 in.
Hertz 60Hz
Lighting Fluorescent
Voltage 115V
Depth (English) 37 in.
Width (Metric) 152.4cm
Depth (English) Interior 30 in.
Height (English) 83 in.
Depth (Metric) Interior 76.2cm
Material Stainless Steel
Type Fume Hood
Height (English) Interior 32 in.
Height (Metric) 210.8cm
Includes Left Access Opening
Ports 2 Utility
Weight 965 lb.
Compliances SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE
Electrical Requirements 115V 60Hz
Sash Opening (English) 12 in.
Depth (Metric) 93.98cm