Please login to your online account to display your discounted pricing

Labconco™ XPert™ Bulk Powder Filtered Systems


Protects from hazardous airborne particulates and fumes generated during powder transfer operations

Manufacturer: labconco™  LCN3961602

 View more versions of this product

Catalog No. 10-304-155

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Height (English) Interior 32 in.
Lighting Fluorescent
Type Fume Hood
Depth (Metric) Interior 76.2cm
Voltage 115V
Ports 2 Utility
Hertz 60Hz
Compliances SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE
Height (English) 83 in.
Includes Right Access Opening
Sash Opening (English) 12 in.
Height (Metric) 210.8cm
Weight 1105 lb.
Depth (English) Interior 30 in.
Electrical Requirements 115V 60Hz
Depth (Metric) 93.98cm
Width (English) 72 in.
Width (Metric) 182.8cm
Depth (English) 37 in.
Material Stainless Steel
Sash Opening (Metric) 30.5cm