missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ XPert™ Bulk Powder Filtered Stations

Designed to protect the user during powder transfer operations

Manufacturer:  Labconco™ LCN3962600

 View more versions of this product

Catalog No. 10-265-152

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

115V 60Hz
12 in.
60Hz
115V
93.98cm
30 in.
76.2cm
Bulk Powder Filtered Station
220.4cm
2 Utility
69.6 in.
SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE
30.5cm
72 in.
Fluorescent
37 in.
182.8cm
86.8 in.
Stainless Steel
32 in.
Left Side Access Opening
1075 lb.
176.8cm