Please login to your online account to display your discounted pricing

Labconco™ XPert™ Bulk Powder Enclosures


Exhausts hazardous airborne particulates and fumes to outside

Manufacturer: labconco™  LCN3963600

 View more versions of this product

Venting; can be connected to a remote blower or house exhaust and exhausted to the outside.

Catalog No. 10-265-160

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Compliances SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE
Height (English) Interior 32 in.
Height (English) 75.5 in.
Material Stainless Steel
Depth (Metric) 93.98cm
Depth (English) Interior 30 in.
Width (English) Interior 70.8 in.
Width (Metric) Interior 179.8 cm
Electrical Requirements 115V 60Hz
Depth (Metric) Interior 76.2cm
Weight 1000 lb.
Height (Metric) 191.7cm
Ports 2 Utility
Type Enclosure, with access opening left side
Hertz 60Hz
Voltage 115V
Lighting Fluorescent
Sash Opening (Metric) 30.5cm
Sash Opening (English) 12 in.
Depth (English) 37 in.
Width (Metric) 182.8cm
Width (English) 72 in.