missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ XPert™ Bulk Powder Enclosures

Exhausts hazardous airborne particulates and fumes to outside

Manufacturer:  Labconco™ LCN3963500

 View more versions of this product

Catalog No. 10-265-156

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Requires : Venting; can be connected to a remote blower or house exhaust and exhausted to the outside.

Specifications Description & Specifications

Specifications

115V 60Hz
12 in.
60Hz
115V
93.98cm
30 in.
76.2cm
Enclosure, with access opening left side
191.7cm
870 lb.
149.4cm
30.5cm
60 in.
Fluorescent
37 in.
152.4cm
75.5 in.
Stainless Steel
32 in.
2 Utility
58.8 in.
SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE