missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Washer Inserts

For use with Labconco FlaskScrubber™ and SteamScrubber™ Washers

Manufacturer:  Labconco™ LCN4595200

 View more versions of this product

Catalog No. 10-359-123


 

Specifications Description & Specifications

Specifications

16 x 1 to 50mL pipet
Two 1.25 in. mounting spindles and PVC coated holder
Each
Pipet Insert
Spindle
0.9kg
Type 304 Stainless Steel
Used for direct injection cleaning of pipets
2 lb.
FlaskScrubber™, FlaskScrubber™ Vantage series, SteamScrubber™ glassware washer