Labconco™ Washer Inserts
For use with Labconco FlaskScrubber™ and SteamScrubber™ Washers
Manufacturer:  Labconco™ LCN4591601
 View more versions of this product
Catalog No. 10-359-122
Specifications Description & Specifications
Specifications
Used to hold variety of labware
5.2 in.
19.8cm
Spindle, Standard
48-Pin Insert
0.9kg
7.8 in.
48-Pins accommodate variety of labware
49cm
FlaskScrubber™, SteamScrubber™ glassware washer
2 lb.
Type 304 Stainless Steel, Plastic tip
Each
13.2cm
Lower and Upper racks
48.0
19.3 in.