missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Telescoping Base Stands

For 4 ft. telescoping base stand

Manufacturer:  Labconco™ LCN3811103

 View more versions of this product

Catalog No. 10-269-306


 

Specifications Description & Specifications

Specifications

9.9D x 60 in. W
Base stand
Epoxy Coated Steel
25.14cm
9.9 in.
60 in.
Hook on the lower rear horizontal cross member of the telescoping base stand
5 ft. telescoping base stand
152.4cm