Labconco™ SpillStopper™ Work Surfaces for 5ft. Protector™Premier™Hoods
Keep your equipment safe from spills with the Labconco SpillStopper Work Surface Protector - specifically dished and contoured to conform to the interior liner of Protector Laboratory Hoods.
Supplier: Labconco™ 9500500
Description
SpillStopper work surfaces are dished to contain spills
- The front edge has large radius to aerodynamically direct airflow in hood
- Some surfaces have cutouts for surface-mounted service fixture and/or 3 x 6 in. cup sink
- Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
- Service fixture and cup sink are sold separately
Specifications
No Cutouts | |
Epoxy Resin |