Labconco™ SpillStopper™ Work Surfaces for 4ft. Protector™XStream
SpillStopper work surfaces are dished to contain spills
Supplier: Labconco™ 9503400
Description
- Front edge has large radius to aerodynamically direct airflow in the hood
- Some surfaces have cutouts for a surface-mounted service fixture and/or a 3 x 6 in. cup sink
- Flat work surfaces may also be coupled with an adjustable height base stand to provide auxiliary workspace
- Service fixture and cup sink are sold separately
Specifications
No Cutouts | |
Epoxy Resin |