Please login to your online account to display your discounted pricing

Labconco™ Shelves for FreeZone™ Freeze-Dry Systems


Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications

Manufacturer: labconco™  LCN7442100

 View more versions of this product

Catalog No. 10-030-152

  • / Each
 

Description & Specifications

Specifications

Diameter (English) 6.25 in.
Diameter (Metric) 15.87cm
For Use With (Application) Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
For Use With (Equipment) Freezone™ drying chambers
Height (English) 9.4 in.
Height (Metric) 23.87cm
Includes Three aluminum shelves with 0.25 in. rims, three stainless steel support rods, nine clips for shelf height adjustment, and three rubber feet
Size Small
Weight (English) 1.5 lb.
Weight (Metric) 0.7kg