Please login to your online account to display your discounted pricing
Labconco™ Shelves for FreeZone™ Freeze-Dry Systems
Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
$1,209.63 - $1,341.29
Specifications
For Use With (Application) Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
For Use With (Equipment) Freezone™ drying chambers
Includes Three aluminum shelves with 0.25 in. rims, three stainless steel support rods, nine clips for shelf height adjustment, and three rubber feet
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Height (English) Size Weight (English) Price Quantity    
10-030-152
labconco™
LCN7442100
6.25 in. 9.4 in. Small 1.5 lb. Each for $1,209.63
10-030-153
labconco™
LCN7441700
10 in. 12.2 in. Large 2 lb. Each for $1,341.29
Description & Specifications
Specifications
For Use With (Application) Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
For Use With (Equipment) Freezone™ drying chambers
Includes Three aluminum shelves with 0.25 in. rims, three stainless steel support rods, nine clips for shelf height adjustment, and three rubber feet
Three 6.5 in. (16.5cm), 7.25 in. (18.4cm), or 10 in. (25.4cm) aluminum shelves with 1/4 in. (0.6cm) rims, three stainless-steel support rods, nine clips for adjustment, and three rubber feet.
Also available: Heated shelves that include ambient to +40°C (+104°F) variable heat control (contact your Fisher Customer Service Representative for details).