missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ ScrubberMate™ Glassware and Rack Cart
Provides convenient storage and transport of extra Labconco washer racks, inserts or miscellaneous glassware
Manufacturer: Labconco™ LCN8027000
Description
- Outfitted with 2.5 in. dia. (6.4cm) casters with toe locks
- L x W x H: 23.3 x 22.6 x 31.6 in. (56.6 x 57.4 x 80.3cm)
Specifications
31.6 in. | |
150 lb. | |
HDPE (High-density Polyethylene) | |
23.3 in. | |
57.4cm | |
Provides convenient storage and transport of extra Labconco up to 2 washer racks, 4 wire inserts or miscellaneous Glassware | |
70.1 lb. |
22.6 in. | |
31.8kg | |
80.3cm | |
59.2cm | |
23.3 x 22.6 x 31.6 in. (59.2 x 57.4 x 80.3cm) | |
2.5 in. dia. Casters with toe locks | |
31.8kg |